missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD9 Human ProcartaPlex™ Simplex Kit

Product Code. 17430733
Change view
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
Product Code. Quantity unitSize
17430733 96 Tests 96 tests
1 options
This item is not returnable. View return policy

Product Code. 17430733

Brand: Invitrogen™ EPX01012434901

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The Human CD9 Simplex ProcartaPlex Kit measures CD9 protein and is designed to be combinable with other Simplex kits so that users can create their own multiplex panel that utilizes Luminex xMAP technology for protein detection/quantitation.

  • The Human CD9 Simplex ProcartaPlex Kit measures CD9 protein
  • Designed to be combinable with other Simplex kits so that users can create their own multiplex panel that utilizes Luminex xMAP technology for protein detection/quantitation
  • When combining multiple Simplex kits (e.g., when not using a pre-configured multiplex panel), only one buffer kit (EPX010-10420-901) is needed for each assay plate regardless of plex size

About ProcartaPlex assays for the Luminex platform

  • ProcartaPlex immunoassays are based on the principles of a sandwich ELISA
  • They use two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument
  • ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant
  • Analyzed results can be obtained in just four hours
  • Validated as a panel to the highest industry standard, including protein target combinability and cross-reactivity testing
  • Panels are available in multiple formats across six species (human, mouse, rat, nonhuman primate, porcine, and canine)
  • Luminex technology is the most referenced multiplexing platform for protein detection and quantitation
TRUSTED_SUSTAINABILITY

Specifications

Product Type Simplex Kit
Quantity 96 Tests
Species Human
Combinability Combinable
Bead Region 19
Assay Sensitivity See Certificate of Analysis
Assay Range See Certificate of Analysis
Format Simplex Kit
For Use With (Equipment) Luminex Instruments
Gene CD9 molecule
Gene Alias MIC3, MRP-1, BTCC-1, DRAP-27, TSPAN29, TSPAN-29, 5H9 antigen
Gene ID (Entrez) 928
Gene Symbol CD9
Interassay CV Less than 15%
Intraassay CV Less than 15%
Product Line ProcartaPlex
Protein CD9 antigen
Sample Type Serum, Plasma, Cell Culture Supernatants
Sample Volume Serum, Plasma: 25 μL; CCS: 50 μL
UniProt ID P21926
Content And Storage

• 1 vial Capture Beads (50X)
• 1 vial Biotinylated Detection Antibody (50X)
• 2 vials Human Standard Mix 11 (lyophilized)

Store at 2°C to 8°C.

Research Area Oncology
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.