missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD9 Antibody (MEM-61), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-327JF646
This item is not returnable.
View return policy
Description
CD9 Monoclonal specifically detects CD9 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CD9 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CD9 | |
| Pre-B cell line NALM-6 | |
| 0.1 mL | |
| Primary | |
| The antibody MEM-61 recognizes an epitope on second extracellular domain (EC2) of CD9 antigen, a 24 kDa transmembrane protein expressed on platelets, monocytes, pre-B lymphocytes, granulocytes and activated T lymphocytes. HLDA VI; WS Code P P-15 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MEM-61 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| BA2, BA-2/p24 antigen, BTCC-1, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24) | |
| Mouse | |
| Protein A purified | |
| Cancer, Embryonic Stem Cell Markers, Stem Cell Markers | |
| 928 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction