missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD9 Monoclonal antibody specifically detects CD9 in Human, Bovine, Canine, Equine, Rabbit, Sheep samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | CD9 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | HI9a |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 0.5 ug/10 million cells in 100 ul |
| Formulation | PBS, pH 7.2 |
| Gene Alias | BA2, BA-2/p24 antigen, BTCC-1, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24) |
| Host Species | Mouse |
| Immunogen | A recombinant protein of human CD9 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?