missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD9 Antibody (Genentech patent anti-CD9) [Janelia Fluor™ 669], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28088JF669
This item is not returnable.
View return policy
Description
CD9 Monoclonal antibody specifically detects CD9 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CD9 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| BA2, BA-2/p24 antigen, BTCC-1, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24) | |
| CD9 | |
| 0.1 mL | |
| Cancer, Stem Cell Markers | |
| 928 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Genentech patent anti-CD9 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction