missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CD9 Antibody (Genentech patent anti-CD9) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28088-50ug
This item is not returnable.
View return policy
Description
CD9 Monoclonal antibody specifically detects CD9 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CD9 | |
| Monoclonal | |
| Unconjugated | |
| P21926 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Genentech patent anti-CD9 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| BA2, BA-2/p24 antigen, BTCC-1, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24) | |
| CD9 | |
| 50 μg | |
| Cancer, Stem Cell Markers | |
| 928 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction