missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD8b Monoclonal Antibody (SIDI8BEE), NovaFluor™ Blue 585, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£154.00 - £340.00
Specifications
| Antigen | CD8b |
|---|---|
| Clone | SIDI8BEE |
| Concentration | 0.8 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055662
|
Invitrogen™
H057T03B04-A |
100 Tests |
£340.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30055011
|
Invitrogen™
H057T02B04-A |
25 Tests |
£154.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD8b Monoclonal antibody specifically detects CD8b in Human, Non-human Primate samples. It is validated for Flow CytometrySpecifications
| CD8b | |
| 0.8 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human, Monkey | |
| P10966 | |
| 926 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| SIDI8BEE | |
| Flow Cytometry | |
| NovaFluor Blue 585 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| CD8; CD8 antigen 37 kDa chain; CD8 antigen beta polypeptide; CD8 antigen beta polypeptide precursor; CD8 antigen beta-chain; CD8 antigen, beta chain; CD8 antigen, beta chain 1; CD8 antigen, beta polypeptide 1 (p37); CD8 antigen, beta-chain; CD8 beta; CD8 beta chain; CD8a; CD8alpha; CD8B; CD8b antigen; CD8b molecule; CD8b molecule pseudogene; CD8B1; CD8beta; CD8BP; LEU2; Leu-2; LY3; Ly-3; Ly-C; Lymphocyte antigen 3; Lyt3; Lyt-3; MAL; OX-8 membrane antigen; p32; P37; T lymphocyte surface glycoprotein beta chain; T-cell membrane glycoprotein Ly-3; T-cell surface glycoprotein; T-cell surface glycoprotein CD8 beta chain; T-cell surface glycoprotein Lyt-3 | |
| IgG1 κ | |
| Affinity chromatography | |
| Cd8b |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title