Learn More
CD8b Monoclonal Antibody (eBioH35-17.2 (H35-17.2)), PE-Cyanine7, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£78.00 - £222.00
Specifications
Antigen | CD8b |
---|---|
Clone | eBioH35-17.2 (H35-17.2) |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15340890
|
Affymetrix eBioscience
25-0083-82 |
100 μg |
£222.00
100µg |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15330890
|
Affymetrix eBioscience
25-0083-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The eBioH35-17.2 monoclonal antibody reacts with the mouse CD8 beta molecule. The CD8 beta chain associates with the CD8 alpha chain to form the CD8 alpha/beta heterodimer expressed on the surface of a majority of thymocytes, and on peripheral cytotoxic alpha beta TCR T cells. CD8 binds to MHC class I and plays a role in T cell development and activation of mature T cells. Applications Reported: This H35-17.2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBioH35-17.2 (H35-17.2) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation.
CD8b Monoclonal antibody specifically detects CD8b in Mouse samples. It is validated for Flow Cytometry.Specifications
CD8b | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
P10300 | |
Cd8b1 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Cd8b1 |
eBioH35-17.2 (H35-17.2) | |
Flow Cytometry | |
PE-Cyanine7 | |
Rat | |
Mouse | |
12526 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.