missing translation for 'onlineSavingsMsg'
Learn More

CD8a Mouse, PE, Clone: RPA-T8, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB19-PE-500Tests

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15951618

  • £467.00 / 500 tests
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD8a
Monoclonal
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
CD8A
Protein A/G Purified
RUO
925
IgG1 κ
Flow Cytometry
RPA-T8
PE
NP_741969
Mouse
500 Tests
Primary
Store at 2-4°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers