missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8A Mouse, Clone: C8/468, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 925-MSM1-P1
This item is not returnable.
View return policy
Specifications
| CD8A | |
| Cell surface | |
| C8/468 | |
| Unconjugated | |
| NP_741969 | |
| Mouse | |
| 32kDa | |
| Protein A/G Purified | |
| RUO | |
| 925 | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide | |
| CD8A | |
| Human CD8 recombinant protein | |
| HuT78 or hPBL. Tonsil | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction