Learn More
CD8A Mouse anti-Human, FITC, Clone: RPA-T8, Abnova™
Mouse monoclonal antibody raised against CD8A.
Brand: Abnova MAB12213.100Reactions
Additional Details : Weight : 0.02000kg
Description
The CD8 antigen is a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediates efficient cell-cell interactions within the immune system. The CD8 antigen acts as a corepressor with the T-cell receptor on the T lymphocyte to recognize antigens displayed by an antigen presenting cell (APC) in the context of class I MHC molecules. The coreceptor functions as either a homodimer composed of two alpha chains, or as a heterodimer composed of one alpha and one beta chain. Both alpha and beta chains share significant homology to immunoglobulin variable light chains. This gene encodes the CD8 alpha chain isoforms. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Specifications
CD8A | |
Monoclonal | |
FITC | |
Flow Cytometry The optimal working dilution should be determined by the end user. | |
CD8/Leu2/MAL/p32 | |
Mouse | |
Affinity Chromatography | |
RUO | |
925 | |
Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Flow Cytometry | |
RPA-T8 | |
Mouse monoclonal antibody raised against CD8A. | |
In PBS, pH7.2 (0.09% Sodium azide, may contain carrier protein/stabilizer). | |
CD8A | |
CD8A | |
100 Reactions | |
Primary | |
Human | |
Liquid |