Learn More
Invitrogen™ CD89 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143971
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human HL-60 whole cell, human U-937 whole cell, human Jurkat whole cell. ICC/IF: Hela cell. Flow: Hela cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
This gene is a member of the immunoglobulin gene superfamily and encodes a receptor for the Fc region of IgA. The receptor is a transmembrane glycoprotein present on the surface of myeloid lineage cells such as neutrophils, monocytes, macrophages, and eosinophils, where it mediates immunologic responses to pathogens. It interacts with IgA-opsonized targets and triggers several immunologic defense processes, including phagocytosis, antibody-dependent cell-mediated cytotoxicity, and stimulation of the release of inflammatory mediators. Multiple alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications
| CD89 | |
| Polyclonal | |
| Unconjugated | |
| Fcar | |
| CD89; CTB-61M7.2; Fc alpha receptor; Fc fragment of IgA receptor; Fc fragment of IgA, receptor for; FcalphaRI; FCAR; IgA Fc receptor; immunoglobulin A Fc receptor; immunoglobulin A Fc receptor transcript variant 1; immunoglobulin A Fc receptor transcript variant 3; immunoglobulin A Fc receptor transcript variant 7; immunoglobulin A Fc receptor transcript variant 9; immunoglobulin A Fc receptor transcript variant deltaEC1; immunoglobulin A Fc receptor transcript variant deltaEC1 delta TM/C; immunoglobulin alpha Fc receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2204 | |
| -20°C | |
| Lyophilized |
| ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| P24071 | |
| Fcar | |
| E.coli-derived human CD89/FCAR recombinant protein (Position: Q22-K287). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.