missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD89 Monoclonal Antibody (A59), PE

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA528547
This item is not returnable.
View return policy
Description
This antibody recognizes an extracellular epitope of CD89, a 55-100 kDa glycoprotein serving as a receptor for IgA and expressed mainly on granulocytes, monocytes and macrophages. Recommended dilution is for 100 μL of whole blood or 1x10^6 cells in suspension.
This gene is a member of the immunoglobulin gene superfamily and encodes a receptor for the Fc region of IgA. The receptor is a transmembrane glycoprotein present on the surface of myeloid lineage cells such as neutrophils, monocytes, macrophages, and eosinophils, where it mediates immunologic responses to pathogens. It interacts with IgA-opsonized targets and triggers several immunologic defense processes, including phagocytosis, antibody-dependent cell-mediated cytotoxicity, and stimulation of the release of inflammatory mediators. Multiple alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications
| CD89 | |
| Monoclonal | |
| PE | |
| Fcar | |
| CD89; CTB-61M7.2; Fc alpha receptor; Fc fragment of IgA receptor; Fc fragment of IgA, receptor for; FcalphaRI; FCAR; IgA Fc receptor; immunoglobulin A Fc receptor; immunoglobulin A Fc receptor transcript variant 1; immunoglobulin A Fc receptor transcript variant 3; immunoglobulin A Fc receptor transcript variant 7; immunoglobulin A Fc receptor transcript variant 9; immunoglobulin A Fc receptor transcript variant deltaEC1; immunoglobulin A Fc receptor transcript variant deltaEC1 delta TM/C; immunoglobulin alpha Fc receptor | |
| Mouse | |
| Size-exclusion chromatography | |
| RUO | |
| 102145896, 2204, 664734 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| A59 | |
| PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
| P24071 | |
| Fcar | |
| Ag8.653 myeloma cells. | |
| 100 Tests | |
| Primary | |
| Cynomolgus Monkey, Human, Monkey, Rhesus Macaque | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction