Learn More
Invitrogen™ CD87 (UPAR) Monoclonal Antibody (VIM5), PE
GREENER_CHOICE
Mouse Monoclonal Antibody
Marca:  Invitrogen™ MA528588
Código de producto. 16302854
  • £248.00 / Pack of 100
Este artículo no se puede devolver. Vea la política de devoluciones
Descripción
Descripción
This antibody recognizes CD87 (urokinase plasminogen activator receptor), a 36-68 kDa single-chain GPI-anchored extracellular glycoprotein expressed on granulocytes, monocytes/macrophages, dendritic cells, endothelial cells, fibroblasts and keratinocytes. Recommended dilution is for 100 μL of whole blood or 1x10^6 cells in suspension.
The urokinase-type plasminogen activator receptor is a key molecule in the regulation of cell-surface plasminogen activation and plays an important role in many normal as well as pathologic processes. The human PLAUR cDNA encodes 335 amino acids including a predicted signal peptide of 22 residues and a hydrophobic C-terminal portion. It produces a highly glycosylated protein of about 50 kD in monocytes where it is anchored to the plasma membrane by glycosyl-phosphatidylinositol linkage. PLAUR, also known as UPAR, is directly associated with the carbohydrate-binding domain of SELL in the membrane of neutrophils, an association analogous to that between PLAUR and beta-2 integrins. PLAUR-mediated calcium mobilization is SELL dependent. UPAR mRNA levels correlate with the invasive potential of endometrial carcinomas and show a 33-fold increase in UPAR mRNA levels in advanced clinical stage endometrial tumors compared with normal endometrial tissue. Furthermore, the increase in UPAR mRNA levels correlated linearly with the progression of disease stage. UPAR protein expressioin correlated positively with rate of recurrence and mortality in patients with endometrial cancer. UPAR appears to be a useful prognostic marker for advanced endometrial cancer.
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
CD87 (UPAR)
Monoclonal
0.025 mg/mL
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
Q03405
PLAUR
human myeloid cell line THP-1.
100 Tests
Primary
Human
Antibody
IgG1
Flow Cytometry
VIM5
PE
PLAUR
Cd87; MO3; Monocyte activation antigen Mo3; Par; plasminogen activator urokinase receptor 3; plasminogen activator, urokinase receptor; PLAUR; Plaur3; sCD87; soluble CD87; uPAR; U-PAR; uPAR-2; uPAR-3; u-plasminogen activator receptor form 2; urinary plasminogen activator receptor 2; urinary plasminogen activator receptor 3; URKR; urokinase plasminogen activator receptor; urokinase plasminogen activator surface receptor; urokinase-type plasminogen activator (uPA) receptor; urokinase-type plasminogen activator receptor
Mouse
Size-exclusion chromatography
RUO
5329
4°C, store in dark, DO NOT FREEZE!
Liquid
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Product Certifications
Promociones
Promociones
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado