missing translation for 'onlineSavingsMsg'
Learn More

CD86 Mouse anti-Human, APC, Clone: 2331 (FUN-1), BD

Mouse Monoclonal Antibody

£117.70 - £950.00

Specifications

Antigen CD86
Clone 2331 (FUN-1)
Applications Flow Cytometry
Classification Monoclonal
Conjugate APC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15890629
View Documents
BD Biosciences
560956
25 Tests
ONLINE EXCLUSIVE PRICE
£122.00
£117.70 / Each
Save £4.30
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15848338
View Documents
BD Biosciences
555660
100 Tests
£950.00
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The 2331 (FUN-1) monoclonal antibody specifically recognizes a 75 kDa transmembrane cell surface protein, CD86 (B70/B7-2), expressed primarily on monocytes, dendritic cells and activated B cells. Competitive binding assays demonstrate that, while both 2331 (FUN-1) and IT2.2 (anti-CD86, Cat. No. 555663) antibodies specifically recognize the same molecule, they react with different epitopes. CD86 is the second ligand for CD28 and CTLA-4 and may play an important role in co-stimulation of T cells in primary immune response. The 2331 (FUN-1) antibody blocks the costimulatory activity of CD86 when tested in functional studies.

Specifications

Specifications

CD86
Flow Cytometry
APC
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
IgG1 κ
Affinity Purified
B7.2; B7-2; B-lymphocyte activation antigen B7-2; B70; BU63; CD28LG2; LAB72
2331 (FUN-1)
Monoclonal
Mouse
Human
Human HBL-1 Cell Line
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.