Learn More
CD86, PE-Alexa Fluor 700, clone: BU63, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD8624
Description
This gene encodes a type I membrane protein that is a member of the immunoglobulin superfamily. This protein is expressed by antigen-presenting cells, and it is the ligand for two proteins at the cell surface of T cells, CD28 antigen and cytotoxic T-lymphocyte-associated protein 4. Binding of this protein with CD28 antigen is a costimulatory signal for activation of the T-cell. Binding of this protein with cytotoxic T-lymphocyte-associated protein 4 negatively regulates T-cell activation and diminishes the immune response. Alternative splicing results in two transcript variants encoding different isoforms. Additional transcript variants have been described, but their full-length sequences have not been determined.
Specifications
| CD86, CD86 (B7-2) | |
| Monoclonal | |
| PE-Alexa Fluor 700 | |
| CD86 | |
| B7-2, B7.2, B70, CD28LG2, LAB72, MGC34413, B-lymphocyte activation antigen B7-2, B-lymphocyte antigen B7-2, B7-2 antigen, BU63, CD28 antigen ligand 2 | |
| Mouse | |
| Purified | |
| ASR, ASR/IVD | |
| 942 | |
| Store at 2 - 8 °C. | |
| Liquid |
| Flow Cytometry | |
| BU63 | |
| PBS with BSA, sucrose and 0.1% sodium azide | |
| P42081 | |
| CD86 | |
| B-lymphoblastoid cell line ARH 77 | |
| 0.5 mL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.