missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD85j (ILT2) Monoclonal Antibody (HP-F1), NovaFluor™ Blue 610-70S, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£154.00 - £340.00
Specifications
| Antigen | CD85j (ILT2) |
|---|---|
| Clone | HP-F1 |
| Concentration | 0.6 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30056030
|
Invitrogen™
H043T03B06-A |
100 Tests |
£340.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30054831
|
Invitrogen™
H043T02B06-A |
25 Tests |
£154.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD85j (ILT2) Monoclonal antibody specifically detects CD85j (ILT2) in Human samples. It is validated for Flow CytometrySpecifications
| CD85j (ILT2) | |
| 0.6 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| Q8NHL6 | |
| 10859 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| HP-F1 | |
| Flow Cytometry | |
| NovaFluor Blue 610-70S | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| CD85 antigen-like family member J; CD85j; Ig-like transcript 2; ILT2; ILT-2; Immunoglobulin-like transcript 2; leucocyte Ig-like receptor B1; leukocyte immunoglobulin like receptor B1; leukocyte immunoglobulin-like receptor subfamily B member 1; leukocyte immunoglobulin-like receptor subfamily B member 1 soluble isoform; leukocyte immunoglobulin-like receptor subfamily B member 3-like; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 1; LILRB1; LIR1; LIR-1; LOC100912456; MIR7; MIR-7; Monocyte/macrophage immunoglobulin-like receptor 7; myeloid inhibitory receptor 7; PIRB; PIR-B | |
| IgG1 κ | |
| Affinity chromatography | |
| LILRB1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title