missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD85d (ILT4) Monoclonal Antibody (42D1), NovaFluor™ Red 710, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£136.00 - £302.00
Specifications
| Antigen | CD85d (ILT4) |
|---|---|
| Clone | 42D1 |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
LILRB2 (ILT4) gene is a member of the leukocyte immunoglobulin-like receptor (LIR) family, which is found in a gene cluster at chromosomal region 19q13.4. The encoded protein belongs to the subfamily B class of LIR receptors which contain two or four extracellular immunoglobulin domains, a transmembrane domain, and two to four cytoplasmic immunoreceptor tyrosine-based inhibitory motifs (ITIMs). The receptor is expressed on immune cells where it binds to MHC class I molecules on antigen-presenting cells and transduces a negative signal that inhibits stimulation of an immune response. It is thought to control inflammatory responses and cytotoxicity to help focus the immune response and limit autoreactivity. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| CD85d (ILT4) | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| CD85 antigen-like family member D; CD85D; CD85d antigen; Ig-like transcript 4; ILT4; ILT-4; leucocyte Ig-like receptor B2; leukocyte immunoglobulin like receptor B2; leukocyte immunoglobulin-like receptor subfamily B member 2; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 2; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 3; LILRB2; Lilrb3; LIR2; LIR-2; LIR2CD85D; MIR10; MIR-10; MIR10LILRA6; monocyte/macrophage immunoglobulin-like receptor 10; myeloid inhibitory receptor 10; Neutrophil immunoglobulin-like receptor 1; Nilr1; NILR-1; paired-Ig-like receptor B; Pirb; type 1 one-pass transmembrane Ig-like receptor | |
| LILRB2 | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! | |
| LILRB2 |
| 42D1 | |
| Flow Cytometry | |
| NovaFluor Red 710 | |
| Rat | |
| Human | |
| Q8N423 | |
| 10288 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title