missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD84/SLAMF5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21300-100ul
This item is not returnable.
View return policy
Description
CD84/SLAMF5 Polyclonal antibody specifically detects CD84/SLAMF5 in Human samples. It is validated for Western Blot, Immunofluorescence
Specifications
| CD84/SLAMF5 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| CD84 antigen, CD84 antigen (leukocyte antigen), CD84 molecule, Cell surface antigen MAX.3, DKFZp781E2378, hCD84, hly9-beta, leucocyte differentiation antigen CD84, leukocyte antigen CD84, Leukocyte differentiation antigen CD84, mCD84, Signaling lymphocytic activation molecule 5, SLAM family member 5, SLAMF5LY9B | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: RRQGRIFPEDAASKKTIYTYIMASRNTQPAESRIYDEILQSKVLPSKEEPVNTVYSEVQFADKMGKASTQDSKPPGTS | |
| 100 μg | |
| CD Markers, Immunology, Stem Cells | |
| 8832 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction