missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD84/SLAMF5 Antibody (152-1D5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-44345-0.02MG
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence
CD84/SLAMF5 Monoclonal specifically detects CD84/SLAMF5 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunofluorescence.
Specifications
| CD84/SLAMF5 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml | |
| CD84 antigen, CD84 antigen (leukocyte antigen), CD84 molecule, Cell surface antigen MAX.3, DKFZp781E2378, hCD84, hly9-beta, leucocyte differentiation antigen CD84, leukocyte antigen CD84, Leukocyte differentiation antigen CD84, mCD84, Signaling lymphocytic activation molecule 5, SLAM family member 5, SLAMF5LY9B | |
| Mouse | |
| 74 kDa | |
| 0.02 mg | |
| Primary | |
| Recognizes a protein of 74kDa, identified as CD84 (Workshop V; Code A085, C057). It is expressed on mature B cells and on B-cell lines, including pre-B-cell lines, but not on plasma cell lines. Immunohistochemical studies demonstrated that CD84 strongly expressed on tissues macrophages. CD84 is also highly expressed on platelets and, at low levels on peripheral blood T cells. It is a highly N-glycosylated protein and belongs to immunoglobulin superfamily. It may play a role in leukocyte activation. | |
| Store at 4C. | |
| IgG1 κ |
| Flow Cytometry, SDS-Page, Immunofluorescence | |
| 152-1D5 | |
| Unconjugated | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| CD84 | |
| Spleen cells of a patient with hairy cell leukemia | |
| Protein A or G purified | |
| RUO | |
| 8832 | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion