missing translation for 'onlineSavingsMsg'
Learn More

CD83 Mouse anti-Human, PE, Clone: HB15E, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 556855

 View more versions of this product

Product Code. 15818788

  • ONLINE EXCLUSIVE PRICE
    £278.00
    £265.65 / Each
    Save £12.35
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reacts with a 45kDa transmembrane protein member of the Ig superfamily. CD83 is composed of a single V-type Ig extracellular domain with a C-terminal cytoplasmic tail. CD83 is found mainly on follicular dendritic cells, circulating dendritic cells, interdigitating dendritic cells in lymphoid tissues, in vitro-generated dendritic cells and thymic dendritic cells. However, its expression is not restricted to dendritic cells. CD83 is also expressed on some germinal center B cells and some lymphoblastoid cell lines. Although its function is not known, it may play a role in cell-cell interaction during antigen presentation. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: HB15e
Isotype: IgG1, κ
Species Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD83
Monoclonal
PE
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human CD83 transfected COS cells
100 Tests
Immunology
Human
IgG1 κ
Flow Cytometry
HB15E
BL11; HB15; B-cell activation protein
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.