missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD83 Monoclonal Antibody (K70018_13H7), Invitrogen™
Mouse Monoclonal Antibody
£416.00 - £734.00
Specifications
| Antigen | CD83 |
|---|---|
| Clone | K70018_13H7 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30200327
|
Invitrogen™
MA556275 |
200 μL |
£734.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30203545
|
Invitrogen™
MA555895 |
50 μL |
£416.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: PEVKVACSED VDLPCTAPWD PQVPYTVSWV KLLEGGEERM ETPQEDHLRG QHYHQKGQNG SFDAPNERPY SLKIRNTTSC NSGTYRCTLQ
CD83 Monoclonal antibody specifically detects CD83 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| CD83 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| B-cell activation protein; BL11; CD antigen CD83; CD83; CD83 antigen; CD83 antigen (activated B lymphocytes, immunoglobulin superfamily); CD83 molecule; cell surface protein HB15; cell-surface glycoprotein; HB15; hCD83; mCD83 | |
| Recombinant human CD83 (immunogen range is 21-110). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| CD83 |
| K70018_13H7 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q01151 | |
| 9308 | |
| IgG2a | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title