missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD83 Biosimilar Recombinant Monoclonal Antibody
GREENER_CHOICE

Recombinant Monoclonal Antibody

£397.00 - £1002.00

Specifications

Antigen CD83 Humanized Biosimilar
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30282781
GREENER_CHOICE
View Documents
Invitrogen™
MA559251
100 μg
£397.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30283619
View Documents
Invitrogen™
MA559252
1 mg
£1002.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term Genentech patent anti-CD83.

CD83 cell surface antigen is a 40-45kD glycoprotein expressed by peripheral blood dendritic cells. Peripheral lymphocytes can be induced to express very low levels of CD83 after culture in agents such as Con A or PHA. In immunohistology, CD83 is shown to be expressed strongly by interfollicular interdigitating reticulum cells and more weakly by cells within germinal centres. CD83 is also expressed by Langerhan's cells in the skin. The CD83 antigen is a 186-amino-acid single-chain glycoprotein and this molecule is a member of the immunoglobulin superfamily that is composed of an extracellular V-type Ig-like single domain, a transmembrane region, and a short, 40-amino-acid cytoplasmic tail. CD83 antigen undergoes extensive post-translational glycosylation, since the determined Mr is twice the predicted size of the core protein. However, CD83+ cells have a unique cell surface immuno-phenotype that does not correlate with that of T cells, B cells, NK cells, or cells of the myelomonocytic lineage. CD83+ cells coexpress the highest levels of MHC class II molecules, when compared with other leucocyte lineages. They also co-express T cell markers (CD2, CD5), B cell markers (CD40, CD78), myeloid cell markers (CD13, CD33, CD36), cytokine receptors as well as other cell surface molecules. Diseases associated with CD83 dysfunction include plague and Rift Valley Fever.
Specifications

Specifications

CD83 Humanized Biosimilar
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Lyophilized
IgG2SA
Human
Primary
Protein A
1 mg/mL
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Unconjugated
Human
RUO
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.