missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Erythropoietin/EPO Antibody (EPO/1367), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-53241-20UG
This item is not returnable.
View return policy
Description
Erythropoietin/EPO Monoclonal specifically detects Erythropoietin/EPO in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| Erythropoietin | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 4 - 8 ug/ml, SDS-Page | |
| EP, epoetin, erythropoietin, MGC138142, MVCD2 | |
| Mouse | |
| 37 kDa | |
| 20 μg | |
| Angiogenesis, Cancer, Cytokine Research, HIF Target Genes, Hypoxia | |
| 2056 | |
| Store at 4C. | |
| IgG |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page | |
| EPO/1367 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| EPO | |
| Recombinant fragment of human EPO protein (aa28-162) (exact sequence is proprietary) | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction