missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CD83 Antibody (Genentech patent anti-CD83) - Humanized, IgG2SA, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28087-50ug
This item is not returnable.
View return policy
Description
CD83 Monoclonal antibody specifically detects CD83 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CD83 | |
| Monoclonal | |
| Unconjugated | |
| Q01151 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG2 |
| Flow Cytometry, ELISA, Functional Assay | |
| Genentech patent anti-CD83 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| B-cell activation protein, BL11CD83 antigen, CD83 antigen (activated B lymphocytes, immunoglobulin superfamily), CD83 molecule, Cell surface protein HB15, cell-surface glycoprotein, HB15hCD83 | |
| CD83 | |
| 50 μg | |
| Dendritic Cell Markers | |
| 9308 | |
| Human | |
| Purified |
Product Content Correction
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido