missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD82 Monoclonal Antibody (07)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA529131
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human CD82/KAI-1.
This metastasis suppressor gene product is a membrane glycoprotein that is a member of the transmembrane 4 superfamily. Expression of this gene has been shown to be downregulated in tumor progression of human cancers and can be activated by p53 through a consensus binding sequence in the promoter. Its expression and that of p53 are strongly correlated, and the loss of expression of these two proteins is associated with poor survival for prostate cancer patients. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications
| CD82 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P27701 | |
| CD82 | |
| Recombinant Human CD82/KAI-1 Protein (Gly111-Leu228). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| 07 | |
| Unconjugated | |
| CD82 | |
| 4F9; AA682076; AL023070; C33; C33 antigen; Cd82; CD82 antigen; CD82 antigen (R2 leukocyte antigen antigen detected by monoclonal and antibody IA4)); CD82 antigen (R2 leukocyte antigen, antigen detected by monoclonal and antibody IA4)); CD82 molecule; GR15; IA4; Inducible membrane protein R2; KAI1; Kangai 1 (suppression of tumerigenicity 6 prostate) CD82 antigen; kangai 1 (suppression of tumorigenicity 6), prostate; kangai 1 (suppression of tumorigenicity 6, prostate); Kangai 1 (suppression of tumorigenicity 6, prostate; CD82 antigen (R2 leukocyte antigen, antigen detected by monoclonal and antibody IA4)); metastasis suppressor Kangai-1; Metastasis suppressor Kangai-1 homolog; R2; SAR2; ST6; Suppressor of tumorigenicity 6 protein; Tetraspanin-27; Tspan27; tspan-27 | |
| Mouse | |
| Protein A | |
| RUO | |
| 3732 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction