missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD82/Kai-1 Mouse anti-Human, Clone: TS82b, Novus Biologicals™
Mouse Monoclonal Antibody
£357.00 - £720.00
Specifications
| Antigen | CD82/Kai-1 |
|---|---|
| Clone | TS82b |
| Concentration | 1 mg/ml |
| Dilution | Western Blot, Flow Cytometry, Immunoprecipitation |
| Applications | Western Blot, Flow Cytometry, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18019205
|
Novus Biologicals
NBP3-18118-200UL |
200 μg |
£357.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18365906
|
Novus Biologicals
NBP3-18118-500UL |
500 μg |
£720.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD82/Kai-1 Monoclonal antibody specifically detects CD82/Kai-1 in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunoprecipitationSpecifications
| CD82/Kai-1 | |
| 1 mg/ml | |
| Western Blot, Flow Cytometry, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| PBS. Sterile-filtered through 0.22 um. | |
| 3732 | |
| IgG1 κ | |
| Affinity purified |
| TS82b | |
| Western Blot, Flow Cytometry, Immunoprecipitation | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD82 antigenC33, CD82 molecule, IA4C33 antigen, Inducible membrane protein R2, KAI1GR15, kangai 1 (suppression of tumorigenicity 6, prostate; CD82 antigen (R2 leukocyteantigen, antigen detected by monoclonal and IA4)), Metastasis suppressor Kangai-1, R2, SAR2tetraspanin-27, ST6tspan-27, Suppressor of tumorigenicity 6 protein, Tetraspanin-27, Tspan-27, TSPAN274F9 | |
| Jurkat and HEL cell lines. Hybridoma: Myeloma X63/AG8. x Balb/c spleen cells | |
| Primary | |
| Store at 4C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title