missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD81 Rabbit anti-Human, Mouse, Rat, Clone: C81/2885R, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-07475-100ug
This item is not returnable.
View return policy
Description
CD81 Monoclonal specifically detects CD81 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry-Paraffin.
Specifications
| CD81 | |
| Monoclonal | |
| 0.2 mg/mL | |
| ELISA, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| CD81 antigen, CD81 antigen (target of antiproliferative 1), CD81 molecule, CVID6, TAPA1, TAPA-1, Target of the antiproliferative 1, Tetraspanin-28, Tspan-28, tspan-28,26 kDa cell surface protein TAPA-1, TSPAN28S5.7 | |
| Recombinant full-length human CD81 protein | |
| 100 μg | |
| Adaptive Immunity, Cell Cycle and Replication, Immunology | |
| 975 | |
| Store at 4°C. | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| C81/2885R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction