missing translation for 'onlineSavingsMsg'
Learn More

CD81 Rabbit anti-Human, Mouse, Rat, Clone: 1.3.3.22, Novus Biologicals™

Product Code. 18333012 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
Unit Size:
0.2mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18333012 0.025 mg 25µg
18389293 0.2 mg 0.2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18333012 Supplier Novus Biologicals Supplier No. NBP3120800.025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CD81 Monoclonal antibody specifically detects CD81 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD81
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 1.3.3.22
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias CD81 antigen, CD81 antigen (target of antiproliferative 1), CD81 molecule, CVID6, TAPA1, TAPA-1, Target of the antiproliferative 1, Tetraspanin-28, Tspan-28, tspan-28,26 kDa cell surface protein TAPA-1, TSPAN28S5.7
Host Species Rabbit
Immunogen This antibody was raised by immunizing mice with a B-Cell line derived from a Burkitt lymphoma.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Cycle and Replication, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 975
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.