Learn More
CD81 Mouse anti-Human, FITC, Clone: JS-81, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 551108
Description
JS-81 identifies the newly clustered CD81 antigen, a 26kDa, type III transmembrane protein (also known as TAPA-1) that is involved in cell growth and signal transduction. CD81 has a very broad cellular distribution, being expressed on cells of hematopoiet
Host Species: Mouse
Clone: JS-81
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Specifications
| CD81 | |
| Monoclonal | |
| FITC | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| JS-81 | |
| TAPA-1 | |
| Mouse | |
| 100 Tests | |
| Immunology | |
| Canine, Human, Rabbit | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.