missing translation for 'onlineSavingsMsg'
Learn More

CD81 Mouse anti-Human, APC, Clone: JS-81, BD

Mouse Monoclonal Antibody

£109.23 - £332.38

Specifications

Antigen CD81
Clone JS-81
Applications Flow Cytometry
Classification Monoclonal
Conjugate APC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15831509
View Documents
BD Biosciences
561958
25 Tests
ONLINE EXCLUSIVE PRICE
£114.00
£109.23 / Each
Save £4.77
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15876528
View Documents
BD Biosciences
551112
100 Tests
ONLINE EXCLUSIVE PRICE
£346.00
£332.38 / Each
Save £13.62
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

JS-81 identifies the newly clustered CD81 antigen, a 26 kDa, type III transmembrane protein (also known as TAPA-1) that is involved in cell growth and signal transduction. CD81 has a very broad cellular distribution, being expressed on cells of hematopoietic, neuroectodermal and mesenchymal origin. MAbs to CD81 have been shown to have anti-proliferative effects on different lymphoid cell lines, particularly those derived from large cell lymphomas. Immunoprecipitation studies reveal that CD81 is a component of a multimolecular complex which can include the Leu13 molecule, and the CD19 and CD21 molecules in B cells. MAbs to CD81 are reported to induce homotypic cell aggregation.

Specifications

Specifications

CD81
Flow Cytometry
APC
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
JS-81
Monoclonal
Mouse
Canine, Human, Rabbit
IgG1 κ
Affinity Purified
TAPA-1
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.