Learn More
CD81 Monoclonal Antibody (Eat-2 (EAT2)), Functional Grade, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
Brand: Affymetrix eBioscience 16-0811-025
Description
The Eat-2 (EAT2) monoclonal antibody reacts with mouse CD81, a molecule also known as TAPA-1. CD81 is expressed on CD4^+CD8^+ thymocytes and broadly in the periphery with high level expressed by B cells, NK cells, macrophages and dendritic cells and lower level on resting T cells. Activation of T cells induces upregulation of CD81. CD81 is involved in homotypic adhesion of B and T cells as well as costimulation of T cells. The Eat-2 antibody is reported to cross-react with rat CD81. Applications Reported: The Eat-2 (EAT2) antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunoblotting (WB). Applications Tested: The Eat-2 (EAT2) antibody has been tested by flow cytometric analysis of mouse thymocytes and splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/ μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
CD81 Monoclonal antibody specifically detects CD81 in Mouse samples. It is validated for Control, Flow Cytometry, Functional Assay, Immunoprecipitation, Western Blot
Specifications
| CD81 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.2 | |
| P35762 | |
| Armenian Hamster | |
| 25 mg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| Flow Cytometry, Functional Assay, Immunoprecipitation, Western Blot | |
| Eat-2 (EAT2) | |
| Functional Grade | |
| CD81 | |
| 26 kDa cell surface protein TAPA-1; CD 81 antigen; Cd81; CD81 antigen; CD81 antigen (target of antiproliferative antibody 1); CD81 molecule; CVID6; S5.7; Tapa1; Tapa-1; Target of the antiproliferative antibody 1; Tetraspanin28; tetraspanin-28; TSPAN28; Tspan-28 | |
| Affinity chromatography | |
| RUO | |
| 12520 | |
| 4° C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.