Learn More
CD81 Armenian Hamster, anti-Mouse, CoraLite Plus 555, Clone: EAT2, Proteintech
Armenian Hamster Monoclonal Antibody
Marque: Proteintech CL555-65220-100UG
Description
The CD81 antibody from Proteintech is a armenian hamster monoclonal antibody generated with cd81+ mouse b lymphoma 38c13 of mouse CD81. This antibody recognizes mouse antigen. The CD81 antibody has been validated for the following applications: FC analysis. This antibody is coralite™ plus 555.
Spécification
| CD81 | |
| EAT2 | |
| CoraLite Plus 555 | |
| Armenian Hamster | |
| Primary | |
| Mouse | |
| 150 kDa | |
| IgG |
| Flow Cytometry | |
| 500 μg/mL | |
| Cd81, CD81 antigen, Tapa 1, Tapa1, Tspan28 | |
| 100 μg | |
| 12520 | |
| Store at 2°C to 8°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.