missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD80 Monoclonal Antibody (MEM-233), APC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen A15707
This item is not returnable.
View return policy
Description
CD80 (B7-1) and CD86 (B7-2) are ligands of T cell critical costimulatory molecule CD28 and of an inhibitory receptor CTLA-4 (CD152). The both B7 molecules are expressed on professional antigen-presenting cells and are essential for T cell activation, the both molecules can also substitute for each other in this process. The question what are the differences in CD80 and CD86 competency has not been fully elucidated yet; there are still conflicts in results about their respective roles in initiation or sustaining of the T cell immune response.
Specifications
| CD80 | |
| Monoclonal | |
| APC | |
| CD80 | |
| B7-1, B7.1, Cd28l, Ly-53, Ly53, MIC17, TS/A-1, LAB7, B7.1, CD28LG, CD28LG1 | |
| Mouse | |
| Purified | |
| RUO | |
| 941 | |
| 4° C | |
| Liquid |
| Flow Cytometry | |
| MEM-233 | |
| PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
| P33681 | |
| CD80 | |
| Human CD80. | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction