missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Mouse, V450, Clone: SK1, enQuireBio™
Mouse Monoclonal Antibody
£185.85 - £1627.50
Specifications
| Antigen | CD8 |
|---|---|
| Clone | SK1 |
| Concentration | 5μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15901608
|
EnquireBio
QAB18-V450-25Tests |
25 Tests |
£185.85
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15991598
|
EnquireBio
QAB18-V450-100Tests |
100 Tests |
£390.60
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15911608
|
EnquireBio
QAB18-V450-500Tests |
500 Tests |
£1627.50
500 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| CD8 | |
| 5μL/Test | |
| Monoclonal | |
| Mouse | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
| 925 | |
| IgG1 κ | |
| Protein A/G Purified |
| SK1 | |
| Flow Cytometry | |
| Violet Fluor 450 (V450) | |
| RUO | |
| NP_741969 | |
| CD8A | |
| Primary | |
| Store at 2-4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title