missing translation for 'onlineSavingsMsg'
Learn More

CD8 Mouse anti-Human, FITC, Clone: 3B5, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen MHCD0801

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10130343

  • £172.00 / Each
Estimated Shipment: 01-09-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 

Description

Description

R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients.

CD8 (Cluster of Differentiation 8) is a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediate efficient cell-cell interactions within the immune system. The CD8 antigen acts as a co-receptor with the T-cell receptor on the T lymphocyte to recognize antigens displayed by an antigen presenting cell (APC) in the context of class I MHC molecules. CD8 is found on a T cell subset of normal cytotoxic/suppressor cells which make up approximately 20-35 % of human peripheral blood lymphocytes. The CD8 antigen is also detected on natural killer (NK) cells, subpopulations of peripheral blood null cells, thymocytes and bone marrow cells. The CD8 co-receptor functions as either a homodimer composed of two alpha chains, or as a disulfide-linked heterodimer composed of one alpha and one beta chain. Both alpha and beta chains share significant homology to immunoglobulin variable light chains. The majority of CD8+ T cells express CD8 as a alpha/beta heterodimer. In HIV, the HIV-2 envelope glycoprotein binds CD8 alpha chain (but not the beta chain).
Specifications

Specifications

CD8
Monoclonal
FITC
CD8B
CD8, Leu-2, MAL, p32, CD8a, CD8b, CD8alpha, CD8beta
Mouse
Purified
IVD
925, 926
4° C, store in dark
Liquid
Flow Cytometry
3B5
PBS with 4mg/mL BSA and 0.1% sodium azide
P01732, P10966
CD8A, CD8B
Human CD8
0.5 mL
Primary
Human
Antibody
IgG2a
Videos
SDS
Documents
Certificates
Special Offers

Special Offers