missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Mouse anti-Human, BUV805, Clone: SK1, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 564912
This item is not returnable.
View return policy
Description
CD8 Monoclonal antibody specifically detects CD8 in Baboon, Cynomolgus, Human, Rhesus Ma samples. It is validated for Flow cytometry.
Specifications
| CD8 | |
| Monoclonal | |
| Brilliant Ultraviolet 805 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 100 Tests | |
| Cell Biology | |
| Baboon, Cynomolgus, Human, Rhesus Monkey | |
| Purified |
| Flow Cytometry | |
| SK1 | |
| Vol. Per test: 5μL | |
| CD8α; CD8A; CD8 alpha; Leu2a; MAL; T8; p32 | |
| Human Peripheral Blood T Cells | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV805 under optimum conditions and unconjugated antibody and free BD Horizon BUV805 were removed. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction