missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Mouse, anti-Human, Atlantic Blue, Clone: SK1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech AB-65146-100TESTS
This item is not returnable.
View return policy
Description
The CD8 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD8. This antibody recognizes human antigen. The CD8 antibody has been validated for the following applications: FC analysis. This antibody is atlantic blue™.
Specifications
| CD8 | |
| SK1 | |
| Atlantic Blue | |
| Mouse | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry | |
| 5 μL/Test | |
| CD8, CD8A, CD8a molecule, Leu2, MAL, p32 | |
| 100 Tests | |
| 925 | |
| Store at 2°C to 8°C. Avoid exposure to light. Stable for one year after shipment. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction