Learn More
CD8 Mouse anti-Human, APC-Cy7, Clone: SK1, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 557834
Description
The SK1 monoclonal antibody specifically binds to CD8 alpha (CD8α). CD8α is a type I transmembrane glycoprotein and a member of the immunoglobulin superfamily. CD8α is expressed by the majority of thymocytes, by subpopulations of αβ T cells and γδ T cells and by some NK cells. Cell surface CD8α is expressed either as a disulfide-linked homodimer (CD8αα) or as a heterodimer (CD8αβ) when disulfide-bonded to a CD8 beta chain (CD8β). CD8-positive αβ T cells coexpress both CD8αα homodimers and CD8αβ heterodimers whereas some γδ T cells and NK cells express CD8αα homodimers. CD8 plays important roles in T cell activation and selection. The extracellular IgSF domain of CD8α binds to a non-polymorphic determinant on HLA class I molecules (α3 domain) and enables CD8 to function as a co-receptor with MHC class I-restricted TCR during T cell recognition of antigen. The cytoplasmic domain of CD8α associates with Lck, a Src family protein tyrosine kinase that is involved in intracellular signaling.
Specifications
| CD8 | |
| Monoclonal | |
| APC-Cyanine7 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human Peripheral Blood T Cells | |
| 100 Tests | |
| Immunology | |
| Baboon, Cynomolgus, Human, Rhesus Monkey | |
| IgG1 κ |
| Flow Cytometry | |
| SK1 | |
| CD8α; CD8A; CD8 alpha; Leu2a; MAL; T8; p32 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.