Learn More
CD8 Monoclonal Antibody (3B5), Pacific Orange™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD0830
Code : 50
Additional Details : Weight : 0.50000kg
Description
Our Pacific Orange™ dye is optimally excited by the violet laser, and it is recommended that a 575, 585 or 600 nm band pass filter be used for optimal detection. Pacific Blue™ and Pacific Orange™ dye conjugates can be simultaneously excited at 405 nm and emit at 455 nm and 551 nm, respectively, facilitating two-color analysis. This antibody recognizes the alpha chain alone as well as the alpha/beta heterodimer.
CD8 Monoclonal antibody specifically detects CD8 in Human samples. It is validated for Flow Cytometry.Specifications
CD8 | |
Monoclonal | |
Pacific Orange | |
CD8B | |
BB154331; CD8; CD8 alpha; CD8 alpha chain; CD8 alpha chain precursor; CD8 antigen; CD8 antigen 32 kDa chain; CD8 antigen 37 kDa chain; CD8 antigen alpha polypeptide; CD8 antigen alpha protein; CD8 antigen alpha protein precursor; CD8 antigen alpha-chain; CD8 antigen beta polypeptide; CD8 antigen beta polypeptide precursor; CD8 antigen beta-chain; CD8 antigen, alpha chain; CD8 antigen, alpha polypeptide; CD8 antigen, alpha polypeptide (p32); CD8 antigen, alpha-chain; CD8 antigen, beta chain; CD8 antigen, beta chain 1; CD8 antigen, beta polypeptide; CD8 antigen, beta polypeptide 1 (p37); CD8 antigen, beta-chain; CD8 beta; CD8 beta chain; CD8 beta-2; CD8a; CD8A antigen alpha; CD8a molecule; CD8A; T-cell surface glycoprotein; CD8alpha; CD8B; CD8b antigen; CD8b molecule; CD8b molecule pseudogene; Cd8b1; CD8beta; CD8BP; fCD8; LEU2; Leu-2; Leu2 T-lymphocyte antigen; leu-2a; Ly-2; LY3; Ly-3; Ly-35; Ly-B; Ly-C; Lymphocyte antigen 3; Lyt2; Lyt-2; Lyt-2.1 lymphocyte differentiation antigen (AA at | |
Mouse | |
purified | |
ASR | |
925, 926 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
3B5 | |
PBS with BSA and 0.1% sodium azide | |
P01732, P10966 | |
Cd8a, CD8B | |
Human CD8 | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |