missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 beta Antibody (V45P5C6*G8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£150.00 - £302.00
Specifications
| Antigen | CD8 beta |
|---|---|
| Clone | V45P5C6*G8 |
| Concentration | 1 mg/mL |
| Dilution | ELISA 1:100 - 1:2000 |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18659898
|
Novus Biologicals
NBP2-50153-0.025ml |
0.025 mL |
£150.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18679076
|
Novus Biologicals
NBP2-50153 |
0.1 mL |
£302.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD8 beta Monoclonal antibody specifically detects CD8 beta in Fish samples. It is validated for ELISASpecifications
| CD8 beta | |
| 1 mg/mL | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Markers | |
| PBS | |
| 926 | |
| IgG1 κ | |
| Protein A purified |
| V45P5C6*G8 | |
| ELISA 1:100 - 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Fish | |
| CD8 antigen, beta polypeptide 1 (p37), CD8b antigen, CD8b molecule, CD8B1P37, LEU2, LY3, LYT3, MGC119115, T lymphocyte surface glycoprotein beta chain, T-cell surface glycoprotein CD8 beta chain | |
| Ovalbumin-conjugated synthetic peptide | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title