missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Antibody (YTS 105.18), Biotin - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-52659B
This item is not returnable.
View return policy
Description
CD8 Monoclonal antibody specifically detects CD8 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen), CyTOF, Neutralization
Specifications
| CD8 | |
| Monoclonal | |
| Biotin | |
| CD_antigen: CD8a, CD8, CD8 antigen, alpha polypeptide (p32), CD8A, CD8a molecule, LEU2, Leu2 T-lymphocyte antigen, MAL, OKT8 T-cell antigen, p32, T cell co-receptor, T8 T-cell antigen, T-cell antigen Leu2, T-cell surface glycoprotein CD8 alpha chain, T-lymphocyte differentiation antigen T8/Leu-2 | |
| Recombinant monoclonal CD8 Antibody (YTS 105.18) - Chimeric was developed against Ly-2-transfected mouse L cells. | |
| 0.1 mL | |
| Adaptive Immunity, Cancer, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Markers | |
| 925 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen), CyTOF | |
| YTS 105.18 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction