missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CD8 Antibody Pair [HRP]

Product Code. 18658650 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18658650

Brand: Novus Biologicals™ NBP2795745Plates

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Polyclonal/Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes an antibody specific for Mouse CD8.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 46.9 pg/mL
Assay Sensitivity 46.88-3000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes an antibody specific for Mouse CD8.
Target Species Mouse
For Use With (Application) Sandwich ELISA
Gene Alias CD8, CD8a molecule, LEU2, p32
Gene ID (Entrez) 925
Gene Symbol CD8A
Host Species Rabbit
Kit Contents
  • Standard: Each vial contains 40 ng of recombinant mouse CD8.
  • Capture Antibody: 0.5 mg/mL of rabbit anti-mouse CD8 monoclonal antibody.
  • Detection Antibody: 0.4 mg/mL rabbit anti-CD8 polyclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Quantity 5 Plates
    Regulatory Status RUO
    Research Discipline Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Signal Transduction, Stem Cell Markers
    Storage Requirements Storage is content dependent.
    Target CD8
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 ug/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20°C to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 3 ng/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.