missing translation for 'onlineSavingsMsg'
Learn More

CD8 Antibody (OX-8), DyLight 405, Novus Biologicals™

Product Code. 18422965 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18422965 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18422965 Supplier Novus Biologicals Supplier No. NBP212523V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD8 Monoclonal antibody specifically detects CD8 in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF, Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD8
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OX-8
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias CD_antigen: CD8a, CD8, CD8 antigen, alpha polypeptide (p32), CD8A, CD8a molecule, LEU2, Leu2 T-lymphocyte antigen, MAL, OKT8 T-cell antigen, p32, T cell co-receptor, T8 T-cell antigen, T-cell antigen Leu2, T-cell surface glycoprotein CD8 alpha chain, T-lymphocyte differentiation antigen T8/Leu-2
Host Species Mouse
Immunogen CD8 Antibody (OX-8) was developed against high Mw glycoproteins from rat thymocytes
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 925
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.