missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD8 Monoclonal antibody specifically detects CD8 in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF, Neutralization
Specifications
Specifications
| Antigen | CD8 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF, Neutralization |
| Classification | Monoclonal |
| Clone | OX-8 |
| Conjugate | Biotin |
| Formulation | PBS |
| Gene Alias | CD_antigen: CD8a, CD8, CD8 antigen, alpha polypeptide (p32), CD8A, CD8a molecule, LEU2, Leu2 T-lymphocyte antigen, MAL, OKT8 T-cell antigen, p32, T cell co-receptor, T8 T-cell antigen, T-cell antigen Leu2, T-cell surface glycoprotein CD8 alpha chain, T-lymphocyte differentiation antigen T8/Leu-2 |
| Host Species | Mouse |
| Immunogen | CD8 Antibody (OX-8) was developed against high Mw glycoproteins from rat thymocytes |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?