missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Antibody (76-2-11), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-28237FR
This item is not returnable.
View return policy
Description
CD8 Monoclonal specifically detects CD8 in Porcine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| CD8 | |
| Monoclonal | |
| DyLight 680 | |
| CD8, CD8a molecule, LEU2, p32 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Porcine CD8a, Mr 32-34 kDa | |
| Store at 4C in the dark. | |
| IgG2b κ |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 76-2-11 | |
| Flow Cytometry | |
| CD8A | |
| Fresh dd miniature swine thymocytes | |
| 0.25 mL | |
| Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Signal Transduction, Stem Cell Markers | |
| 925 | |
| Pig | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction