missing translation for 'onlineSavingsMsg'
Learn More

CD79B Rabbit anti-Human, Clone: IGB/2940R, Novus Biologicals™

Product Code. 18338773 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18338773 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18338773 Supplier Novus Biologicals Supplier No. NBP30744020UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CD79B Monoclonal specifically detects CD79B in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD79B
Applications ELISA
Classification Monoclonal
Clone IGB/2940R
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution ELISA 2-4 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Alias B29B-cell-specific glycoprotein B29, B-cell antigen receptor complex-associated protein beta chain, CD79b antigen, CD79B antigen (immunoglobulin-associated beta), CD79b molecule, immunoglobulin-associated beta, IGBAGM6, Ig-beta, Immunoglobulin-associated B29 protein
Host Species Rabbit
Immunogen Recombinant human CD79B protein fragment (around aa29-159) (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 974
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.