missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD79b Polyclonal Antibody
GREENER_CHOICE

Product Code. 16359344
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16359344 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16359344 Supplier Invitrogen™ Supplier No. PA582382

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: DKDDSKAGME EDHTYEGLDI DQTATYEDIV TLRTGEVKWS VGEHPGQE Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 98%.

CD79b is the B cell antigen receptor Ig beta chain (BCR-Ig beta), which associates with B cell antigen receptor Ig alpha chain to form a functional heterodimer that interacts with membrane immunoglobulin (mIg). The mIgM component binds antigen and the Ig alpha /I g beta heterodimer tranduces signals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD79b
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Cd79b
Gene Accession No. P40259
Gene Alias AGM6; B29; B-cell antigen receptor complex-associated protein beta chain; B-cell-specific glycoprotein B29; CD79; CD79b; CD79B antigen; CD79b molecule; Cd79b molecule, immunoglobulin-associated beta; Ig beta; IGB; Igbeta; Ig-beta; Immunoglobulin-associated B29 protein; immunoglobulin-associated beta; RP23-418O11.7
Gene Symbols Cd79b
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CD79B. Recombinant protein control fragment (Product #RP-89949).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 974
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.