Learn More
CD79b Mouse anti-Human, PE, Clone: CB3-1, BD
Mouse Monoclonal Antibody
£189.00 - £192.09
Specifications
| Antigen | CD79b |
|---|---|
| Clone | CB3-1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15881489
|
BD Biosciences
561943 |
25 Tests | |||||||
|
15828358
|
BD Biosciences
555679 |
100 Tests |
£201.00 £192.09 / Each Save £8.91 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunoglobulin (Ig) antigen receptors are composed of a non-covalently- associated complex of Ig and two other proteins, Igα and Igβ, which have been designated in the Fifth International Leukocyte Workshop as CD79a and CD79b respectively. CB3-1 reacts with CD79b, which is expressed on surface Ig(sIg)-positive lymphocytes and B-cell lines but only in the cytoplasm of sIg-negative cells including most terminal deoxynucleotidyl transferase (TdT) positive early pre-B and all cytoplasmic µ positive pre-B cell lines. Antibodies to CD79b are helpful in delineating signal transduction pathways activated via antibody receptors during different stages of B-cell differentiation.
Specifications
| CD79b | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| CB3-1 | |
| Monoclonal | |
| Mouse | |
| Human | |
| IgG1 κ | |
| Affinity Purified | |
| Igβ |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.