missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD79B Mouse anti-Human, Clone: rIGB/1842, Novus Biologicals™
Mouse Monoclonal Antibody
£206.00
Specifications
| Antigen | CD79B |
|---|---|
| Clone | rIGB/1842 |
| Concentration | 0.2 mg/ml |
| Dilution | ELISA 2-4 ug/ml |
| Applications | ELISA |
Description
CD79B Monoclonal specifically detects CD79B in Human samples. It is validated for ELISA.Specifications
| CD79B | |
| 0.2 mg/ml | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| B29B-cell-specific glycoprotein B29, B-cell antigen receptor complex-associated protein beta chain, CD79b antigen, CD79B antigen (immunoglobulin-associated beta), CD79b molecule, immunoglobulin-associated beta, IGBAGM6, Ig-beta, Immunoglobulin-associated B29 protein | |
| Recombinant human CD79B protein fragment (around aa29-159) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
| rIGB/1842 | |
| ELISA 2-4 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 974 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title