Learn More
Invitrogen™ CD79b Monoclonal Antibody (CB3-1), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ 14-0793-95
Description
Description: This CB3-1 monoclonal antibody reacts with human CD79b, the B cell antigen receptor Ig-beta chain. Together with the Ig-alpha chain, also known as CD79a, this cell surface molecule forms a heterodimeric complex that associates with membrane immunoglobulin (mIgM). CD79b expression is exclusive to B lymphocytes and B cell lymphomas. Depending on the maturation state of the B cell, CD79b can be expressed either in the cytoplasm or on the cell surface. The CD79 receptor complex triggers numerous signaling pathways to mediate B cell development, maintenance, and activation. Applications Reported: This CB3-1 antibody has been reported for use in flow cytometric analysis and immunoblotting (WB). Applications Tested: This CB3-1 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
CD79b is the B cell antigen receptor Ig beta chain (BCR-Ig beta), which associates with B cell antigen receptor Ig alpha chain to form a functional heterodimer that interacts with membrane immunoglobulin (mIg). The mIgM component binds antigen and the Ig alpha /I g beta heterodimer tranduces signals.
Specifications
| CD79b | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P40259 | |
| Cd79b | |
| Affinity chromatography | |
| RUO | |
| 974 | |
| 4°C | |
| Liquid |
| Flow Cytometry, Western Blot | |
| CB3-1 | |
| Unconjugated | |
| Cd79b | |
| AGM6; B29; B-cell antigen receptor complex-associated protein beta chain; B-cell-specific glycoprotein B29; CD79; CD79b; CD79B antigen; CD79b molecule; Cd79b molecule, immunoglobulin-associated beta; Ig beta; IGB; Igbeta; Ig-beta; Immunoglobulin-associated B29 protein; immunoglobulin-associated beta; RP23-418O11.7 | |
| Mouse | |
| 10 mg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.